Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cholin Acetate

Catalog No. 6701895G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-018-95G 5 g
67-018-950G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 67-018-95G Supplier Sigma Aldrich Supplier No. 6701895G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cholin acetate is a choline-derivative-based ionic liquid (IL), which can efficiently dissolve microcrystalline cellulose. This bioionic liquid can be prepared by reacting choline hydroxide with acetic acid.Cholin acetate in combination with glycerol, a hydrogen-bond donor forms a eutectic ionic liquid mixture, which is less viscous, biodegradable and also shows high lipase compatibility. This eutectic ionic liquid mixture can be a desirable solvent for the enzymatic production of biodiesel.

Specifications

CAS 14586-35-7
Molecular Formula C7H17NO3
Linear Formula C7H17NO3
Quantity 5 g
Synonym 2-Hydroxyethyl-trimethylammonium acetate; COL Acetate
Molecular Weight (g/mol) 163.21 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.