Learn More
Medchemexpress LLC Choline-13C2 chloride | 202190-49-6 | 98.0% | 141.61 | C5H14ClNO | 10 MG
SDP
Catalog No. 5000297981
Encompass_Preferred
Choline-13C2 chloride is a stable isotope-labeled form of choline chloride used as an internal standard and tracer for metabolic, biochemical, and pharmacokinetic studies. It is supplied as a small, high-purity material intended for analytical and research applications.
  • Stable isotope labeled (two 13C atoms) for mass spectrometry and NMR differentiation.
  • Suitable as an internal standard for LC-MS/MS assays.
  • High purity: 98.0%.
  • Pack size intended for analytical use: 10 MG.
  • CAS number 202190-49-6 for unambiguous identification.
Catalog No. 50-002-97981 Supplier Medchemexpress LLC Supplier No. HYB1337S510MG
Choline-13C2 chloride is a stable isotope-labeled form of choline chloride used as an internal standard and tracer for metabolic, biochemical, and pharmacokinetic studies. It is supplied as a small, high-purity material intended for analytical and research applications.
  • Stable isotope labeled (two 13C atoms) for mass spectrometry and NMR differentiation.
  • Suitable as an internal standard for LC-MS/MS assays.
  • High purity: 98.0%.
  • Pack size intended for analytical use: 10 MG.
  • CAS number 202190-49-6 for unambiguous identification.