Learn More
Medchemexpress LLC Choline-13C2 chloride | 202190-49-6 | 98.0% | 5 MG
SDP
Catalog No. 5000411575
Encompass_Preferred
Choline-13C2 chloride is a 13C labeled Choline chloride, an essential nutrient known for activating alpha7 nicotinic receptors and demonstrating analgesic and anti-inflammatory activity. This compound is valuable as a tracer and an internal standard for quantitative analysis in various analytical techniques.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-11575 Supplier Medchemexpress LLC Supplier No. HYB1337S55MG
Choline-13C2 chloride is a 13C labeled Choline chloride, an essential nutrient known for activating alpha7 nicotinic receptors and demonstrating analgesic and anti-inflammatory activity. This compound is valuable as a tracer and an internal standard for quantitative analysis in various analytical techniques.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs