Learn More
Medchemexpress LLC Choline bitartrate | 87-67-2 | MFCD00036332 | 98.0% | 253.25 g/mol | C9H19NO7 | 1 ML
SDP
Catalog No. 5000211173
Encompass_Preferred
Choline bitartrate is a research-grade methyl donor and precursor to the neurotransmitter acetylcholine, supplied as a ready-to-use 10 mM solution (1 mL) in DMSO. It is used in biochemical and cellular studies involving lipid metabolism, one-carbon/methylation pathways, and neurotransmission. CAS 87-67-2; typical purity 98.0%.
  • Prepared 10 mM solution for convenient dosing and reduced prep time.
  • High purity for reproducible experimental results.
  • Supplied in DMSO for compatibility with common assay systems.
  • Useful as a methyl donor in metabolic and epigenetic studies.
  • Acts as a precursor to acetylcholine for neurotransmission research.
Catalog No. 50-002-11173 Supplier Medchemexpress LLC Supplier No. HY10103610MM 1ML
Choline bitartrate is a research-grade methyl donor and precursor to the neurotransmitter acetylcholine, supplied as a ready-to-use 10 mM solution (1 mL) in DMSO. It is used in biochemical and cellular studies involving lipid metabolism, one-carbon/methylation pathways, and neurotransmission. CAS 87-67-2; typical purity 98.0%.
  • Prepared 10 mM solution for convenient dosing and reduced prep time.
  • High purity for reproducible experimental results.
  • Supplied in DMSO for compatibility with common assay systems.
  • Useful as a methyl donor in metabolic and epigenetic studies.
  • Acts as a precursor to acetylcholine for neurotransmission research.