Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Choline bitartrate | 87-67-2 | MFCD00036332 | 98.4% | 253.25 g/mol | C9H19NO7 | 250 MG
SDP

Catalog No. 5000213232
Encompass_Preferred

Choline bitartrate is a high-purity analytical reference material used for method development, calibration, and quality control in biochemical and pharmaceutical research. Supplied in small laboratory pack sizes, it is intended for use as a methyl donor standard and for validating analytical assays.

  • High purity (98.44%) suitable for analytical applications.
  • Supplied with certificate of analysis and safety data sheet.
  • Available in laboratory-friendly pack sizes, including 250 MG.
  • Identifiable by CAS number 87-67-2 and formula C9H19NO7.
  • Suitable as a methyl donor standard for biochemical assays.

Catalog No. 50-002-13232 Supplier Medchemexpress LLC Supplier No. HY101036R250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choline bitartrate is a high-purity analytical reference material used for method development, calibration, and quality control in biochemical and pharmaceutical research. Supplied in small laboratory pack sizes, it is intended for use as a methyl donor standard and for validating analytical assays.

  • High purity (98.44%) suitable for analytical applications.
  • Supplied with certificate of analysis and safety data sheet.
  • Available in laboratory-friendly pack sizes, including 250 MG.
  • Identifiable by CAS number 87-67-2 and formula C9H19NO7.
  • Suitable as a methyl donor standard for biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.