Learn More
Medchemexpress LLC Choline (bitartrate) | 87-67-2 | MFCD31726185 | 98.4% | 253.25 g/mol | C9H19NO7 | 10 MG
SDP
Catalog No. 5000211171
Encompass_Preferred
Choline (bitartrate) is an analytical reference standard of choline supplied as a white to off-white solid and intended for research and analytical applications such as calibration, method development, and quality control. It has a molecular weight of 253.25 g/mol and is provided with supporting documentation for safe and consistent use.
  • High purity suitable for analytical use.
  • White to off-white solid physical form.
  • Available in small quantities for trace-level calibration.
  • Includes certificate of analysis and safety data sheet.
  • Suitable for method development and quality control.
Catalog No. 50-002-11171 Supplier Medchemexpress LLC Supplier No. HY101036R10MG
Choline (bitartrate) is an analytical reference standard of choline supplied as a white to off-white solid and intended for research and analytical applications such as calibration, method development, and quality control. It has a molecular weight of 253.25 g/mol and is provided with supporting documentation for safe and consistent use.
  • High purity suitable for analytical use.
  • White to off-white solid physical form.
  • Available in small quantities for trace-level calibration.
  • Includes certificate of analysis and safety data sheet.
  • Suitable for method development and quality control.