Learn More
Medchemexpress LLC Choline bitartrate | 87-67-2 | MFCD31726185 | 98.4% | 253.25 g/mol | C9H19NO7 | 25 MG
SDP
Catalog No. 5000212818
Encompass_Preferred
Choline bitartrate is an analytical standard of choline used as a methyl donor and precursor to the neurotransmitter acetylcholine. Supplied as a solid reference material for research and analytical applications, it is suitable for qualitative and quantitative experiments by HPLC, GC, and MS. Typical pack sizes include small-mass standard quantities for assay use.
  • Analytical standard suitable for HPLC, GC, and MS.
  • High purity (98.4%), suitable for quantitative analysis.
  • Solid form ideal for accurate weighing and sample preparation.
  • Available in small standard quantities for assay and method development.
Catalog No. 50-002-12818 Supplier Medchemexpress LLC Supplier No. HY101036R25MG
Choline bitartrate is an analytical standard of choline used as a methyl donor and precursor to the neurotransmitter acetylcholine. Supplied as a solid reference material for research and analytical applications, it is suitable for qualitative and quantitative experiments by HPLC, GC, and MS. Typical pack sizes include small-mass standard quantities for assay use.
  • Analytical standard suitable for HPLC, GC, and MS.
  • High purity (98.4%), suitable for quantitative analysis.
  • Solid form ideal for accurate weighing and sample preparation.
  • Available in small standard quantities for assay and method development.