Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Bitartrate, FCC, 98%, Spectrum™ Chemical
SDP

Catalog No. 18601668 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-668 50 kg Fiber Drum
18-601-665 500 g Poly Bottle
18-601-666 2.5 kg Poly Bottle
18-601-667 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-668 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C122850KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C1228, 87-67-2, C9H19NO7

Spectrum™ Chemical Choline Bitartrate, FCC is an essential nutrient and typically grouped with B-complex vitamins. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 87-67-2
Molecular Formula C9H19NO7
Molecular Weight (g/mol) 253.25
InChI Key QWJSAWXRUVVRLH-LREBCSMRSA-M
IUPAC Name (2-hydroxyethyl)trimethylazanium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
SMILES C[N+](C)(C)CCO.O[C@H]([C@@H](O)C([O-])=O)C(O)=O

Specifications

CAS Max % 100%
Quantity 50 kg
Ignition Residue 0.1%
Optical Rotation +17.5 to +18.5°
Packaging Fiber Drum
Water 0.5%
Percent Purity 98%
Grade FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.