Learn More
Medchemexpress LLC Choline (bitartrate) (standard) | 87-67-2 | MFCD00036332 | 98.4% | 253.25 g/mol | C9H19NO7 | 100 MG
SDP
Catalog No. 5000212022
Encompass_Preferred
Choline (bitartrate) (standard) is an analytical reference standard of choline in its bitartrate salt form, provided as a white crystalline powder for laboratory and analytical use. It functions as a methyl donor and precursor to acetylcholine, and is intended for assay calibration, method development, and quality control in research and analytical laboratories. CAS 87-67-2; molecular weight 253.25 g/mol.
  • High purity (98.44%) confirmed by Q-NMR.
  • Intended for use as an analytical reference standard and assay calibration.
  • White crystalline powder form suitable for accurate weighing and dissolution.
  • Documented molecular formula and molecular weight for traceable characterization.
  • COA and SDS available for batch-specific safety and quality information.
Catalog No. 50-002-12022 Supplier Medchemexpress LLC Supplier No. HY101036R100MG
Choline (bitartrate) (standard) is an analytical reference standard of choline in its bitartrate salt form, provided as a white crystalline powder for laboratory and analytical use. It functions as a methyl donor and precursor to acetylcholine, and is intended for assay calibration, method development, and quality control in research and analytical laboratories. CAS 87-67-2; molecular weight 253.25 g/mol.
  • High purity (98.44%) confirmed by Q-NMR.
  • Intended for use as an analytical reference standard and assay calibration.
  • White crystalline powder form suitable for accurate weighing and dissolution.
  • Documented molecular formula and molecular weight for traceable characterization.
  • COA and SDS available for batch-specific safety and quality information.