Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Choline bromide | 1927-06-6 | MFCD00054244 | 98.0% | 184.07 | C5H14BrNO | 100 G
SDP

Catalog No. 5000282746
Encompass_Preferred

Choline bromide is an organic ammonium salt used as a biochemical assay reagent for laboratory research and analytical applications. It is supplied as a solid with reported purity of 98.0% and is intended for research use only; handle and store according to the supplied safety documentation.

  • Purity 98.0%.
  • Molecular formula C5H14BrNO.
  • Molecular weight 184.07 g/mol.
  • CAS number 1927-06-6.
  • Available as 100 g solid pack.
  • Store sealed, away from moisture; store product at 4°C.

Catalog No. 50-002-82746 Supplier Medchemexpress LLC Supplier No. HYW127656100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choline bromide is an organic ammonium salt used as a biochemical assay reagent for laboratory research and analytical applications. It is supplied as a solid with reported purity of 98.0% and is intended for research use only; handle and store according to the supplied safety documentation.

  • Purity 98.0%.
  • Molecular formula C5H14BrNO.
  • Molecular weight 184.07 g/mol.
  • CAS number 1927-06-6.
  • Available as 100 g solid pack.
  • Store sealed, away from moisture; store product at 4°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.