Learn More
Medchemexpress LLC Choline bromide | 1927-06-6 | MFCD00054244 | 98.0% | 184.07 g/mol | C5H14BrNO | 25 G
SDP
Catalog No. 5000283627
Encompass_Preferred
Choline bromide is the bromide salt of choline used as a biochemical reagent and reference standard in life science research. Supplied as a crystalline, water-soluble compound, it is suitable for use in assays, synthesis, and analytical methods where a quaternary ammonium salt is required.
  • High purity suitable for research use (98.0%).
  • Water-soluble quaternary ammonium compound for biochemical assays.
  • Well-characterized: CAS 1927-06-6, formula C5H14BrNO, MW 184.07 g/mol.
  • Available in small lab-friendly quantities for convenience.
  • Accompanied by datasheet, SDS, and certificate of analysis for traceability.
Catalog No. 50-002-83627 Supplier Medchemexpress LLC Supplier No. HYW12765625G
Choline bromide is the bromide salt of choline used as a biochemical reagent and reference standard in life science research. Supplied as a crystalline, water-soluble compound, it is suitable for use in assays, synthesis, and analytical methods where a quaternary ammonium salt is required.
  • High purity suitable for research use (98.0%).
  • Water-soluble quaternary ammonium compound for biochemical assays.
  • Well-characterized: CAS 1927-06-6, formula C5H14BrNO, MW 184.07 g/mol.
  • Available in small lab-friendly quantities for convenience.
  • Accompanied by datasheet, SDS, and certificate of analysis for traceability.