Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cambridge Isotope Laboratories Choline bromide (D13 98%) 0 1 g

Catalog No. 5000166283
Encompass

Choline bromide (D13 98%) 0 1 g

Catalog No. 50-001-66283 Supplier Cambridge Isotope Laboratories Supplier No. DLM30150.1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choline bromide (D13 98%) 0 1 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.