Learn More
Sigma Aldrich Fine Chemicals Biosciences Choline chloride >=99% | 67-48-1 | MFCD00011721 | 10MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C701710MG
Encompass
Choline chloride >=99% | Purity: >=99% | Mol Wt: 139.62 | 67-48-1 | MFCD00011721 | 10MG
Catalog No. NC1615995