Learn More
Medchemexpress LLC Choline chloride-13C3 | 67-48-1 | 99.5% | 142.60 | C2(13C)3H14ClNO | 10 MG
SDP
Catalog No. 5000297262
Encompass_Preferred
Choline chloride-13C3 is a 13C-labeled stable isotope of choline chloride provided as a small-quantity, high-purity standard for tracer studies and mass spectrometry. It is supplied as the chloride salt of choline with three 13C atoms incorporated and is intended for use as an internal standard, tracer, or labeled reagent in biochemical and analytical workflows.
  • High isotopic labeling for tracer and mass spectrometry applications.
  • High chemical purity suitable for analytical use.
  • Supplied in small pack sizes for research and method development.
  • Water- and DMSO-soluble for easy preparation of standards and solutions.
  • Designed for use as an internal standard and tracer in biochemical assays.
Catalog No. 50-002-97262 Supplier Medchemexpress LLC Supplier No. HYB1337S410MG
Choline chloride-13C3 is a 13C-labeled stable isotope of choline chloride provided as a small-quantity, high-purity standard for tracer studies and mass spectrometry. It is supplied as the chloride salt of choline with three 13C atoms incorporated and is intended for use as an internal standard, tracer, or labeled reagent in biochemical and analytical workflows.
  • High isotopic labeling for tracer and mass spectrometry applications.
  • High chemical purity suitable for analytical use.
  • Supplied in small pack sizes for research and method development.
  • Water- and DMSO-soluble for easy preparation of standards and solutions.
  • Designed for use as an internal standard and tracer in biochemical assays.