Learn More
Medchemexpress LLC Choline Chloride-13C3 | 67-48-1 | 99.7% | 1 MG
SDP
Catalog No. 5000412009
Encompass_Preferred
Choline Chloride-13C3 is the 13C-labeled Choline (chloride). Choline chloride is an organic compound and a quaternary ammonium salt, an acyl group acceptor and choline acetyltransferase substrate. It is also an important additive in feed, especially for chickens, where it accelerates growth.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12009 Supplier Medchemexpress LLC Supplier No. HYB1337S41MG
Choline Chloride-13C3 is the 13C-labeled Choline (chloride). Choline chloride is an organic compound and a quaternary ammonium salt, an acyl group acceptor and choline acetyltransferase substrate. It is also an important additive in feed, especially for chickens, where it accelerates growth.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.