Learn More
Alkali Scientific Choline Chloride, 500 Grams
SDP
Catalog No. 5000660828
Encompass_Preferred
Choline Chloride is a quaternary ammonium salt widely used in cell culture, biochemical research, and nutritional studies. It serves as an essential nutrient involved in membrane structure and lipid metabolism, particularly as a precursor for phosphatidylcholine synthesis. In cell culture applications, choline chloride supports cell growth and viability by contributing to membrane integrity and signaling pathways. It is also used in metabolic research to study neurotransmitter synthesis and methyl group metabolism. The compound is commonly incorporated into media formulations and biochemical assays, providing a stable and water-soluble source of choline for laboratory and research applications.
Catalog No. 50-006-60828 Supplier Alkali Scientific Supplier No. CHO005CP
Choline Chloride is a quaternary ammonium salt widely used in cell culture, biochemical research, and nutritional studies. It serves as an essential nutrient involved in membrane structure and lipid metabolism, particularly as a precursor for phosphatidylcholine synthesis. In cell culture applications, choline chloride supports cell growth and viability by contributing to membrane integrity and signaling pathways. It is also used in metabolic research to study neurotransmitter synthesis and methyl group metabolism. The compound is commonly incorporated into media formulations and biochemical assays, providing a stable and water-soluble source of choline for laboratory and research applications.