Learn More
Medchemexpress LLC Choline (chloride) | 67-48-1 | 99.2% | 100 MG
SDP
Catalog No. 5000412064
Encompass_Preferred
Choline chloride (Standard) is the analytical standard of Choline (chloride) intended for research and analytical applications. It is an essential nutrient that activates alpha7 nicotinic receptors, demonstrating analgesic and anti-inflammatory activity. Glycerophosphoinositol choline can affect diseases such as liver disease, atherosclerosis, and neurological disorders.
  • Used as an analytical standard.
  • Activates alpha7 nicotinic receptors.
  • Possesses analgesic and anti-inflammatory activity.
  • Can affect liver disease, atherosclerosis, and neurological disorders.
  • Suitable for qualitative, quantitative, and methodological research experiments.
  • Applicable in HPLC, GC, and MS.
Catalog No. 50-004-12064 Supplier Medchemexpress LLC Supplier No. HYB1337R100MG
Choline chloride (Standard) is the analytical standard of Choline (chloride) intended for research and analytical applications. It is an essential nutrient that activates alpha7 nicotinic receptors, demonstrating analgesic and anti-inflammatory activity. Glycerophosphoinositol choline can affect diseases such as liver disease, atherosclerosis, and neurological disorders.
  • Used as an analytical standard.
  • Activates alpha7 nicotinic receptors.
  • Possesses analgesic and anti-inflammatory activity.
  • Can affect liver disease, atherosclerosis, and neurological disorders.
  • Suitable for qualitative, quantitative, and methodological research experiments.
  • Applicable in HPLC, GC, and MS.