Learn More
Medchemexpress LLC Choline (chloride) | 67-48-1 | 99.9% | 500 MG
SDP
Catalog No. 5000413243
Encompass_Preferred
Choline chloride is an essential nutrient that activates alpha7 nicotinic receptors and has analgesic and anti-inflammatory activity. Glycerophosphoinositol choline can affect diseases such as liver disease, atherosclerosis, and neurological disorders.
  • Activates alpha7 nicotinic receptors
  • Possesses analgesic and anti-inflammatory activity
  • Influences conditions such as liver disease, atherosclerosis, and neurological disorders
Catalog No. 50-004-13243 Supplier Medchemexpress LLC Supplier No. HYB1337500MG
Choline chloride is an essential nutrient that activates alpha7 nicotinic receptors and has analgesic and anti-inflammatory activity. Glycerophosphoinositol choline can affect diseases such as liver disease, atherosclerosis, and neurological disorders.
  • Activates alpha7 nicotinic receptors
  • Possesses analgesic and anti-inflammatory activity
  • Influences conditions such as liver disease, atherosclerosis, and neurological disorders