Learn More
Medchemexpress LLC Choline chloride | 67-48-1 | MFCD00002821 | >99% | 1 G
SDP
Catalog No. 5000412008
Encompass_Preferred
Choline chloride is an essential nutrient known for activating alpha7 nicotinic receptors and exhibiting analgesic and anti-inflammatory activities. This organic compound, a quaternary ammonium salt, acts as an acyl group acceptor and a substrate for choline acetyltransferase. It plays a role in various conditions, including liver disease, atherosclerosis, and neurological disorders, and is also used as a significant feed additive to promote growth, particularly in chickens.
  • Activates alpha7 nicotinic receptors
  • Exhibits analgesic activity
  • Exhibits anti-inflammatory activity
  • Organic compound and quaternary ammonium salt
  • Acyl group acceptor and choline acetyltransferase substrate
  • Influences liver disease, atherosclerosis, and neurological disorders
  • Promotes growth in animals, especially chickens
Catalog No. 50-004-12008 Supplier Medchemexpress LLC Supplier No. HYB13371G
Choline chloride is an essential nutrient known for activating alpha7 nicotinic receptors and exhibiting analgesic and anti-inflammatory activities. This organic compound, a quaternary ammonium salt, acts as an acyl group acceptor and a substrate for choline acetyltransferase. It plays a role in various conditions, including liver disease, atherosclerosis, and neurological disorders, and is also used as a significant feed additive to promote growth, particularly in chickens.
  • Activates alpha7 nicotinic receptors
  • Exhibits analgesic activity
  • Exhibits anti-inflammatory activity
  • Organic compound and quaternary ammonium salt
  • Acyl group acceptor and choline acetyltransferase substrate
  • Influences liver disease, atherosclerosis, and neurological disorders
  • Promotes growth in animals, especially chickens