Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Choline chloride | 67-48-1 | MFCD00011721 | 99.2% | 10 MG
SDP

Catalog No. 5000297809
Encompass_Preferred

Choline chloride is a quaternary ammonium salt and essential nutrient used as an analytical standard in biochemical and pharmacological research. It is provided as a high-purity crystalline solid suitable for analytical assays, receptor activation studies, and metabolic research.

  • Assay 99.17%.
  • CAS number 67-48-1.
  • Molecular weight 139.62 g/mol.
  • Chemical formula C5H14ClNO.
  • Suitable for in vitro and analytical applications.
  • Available in small reference quantities, such as 10 MG.

Catalog No. 50-002-97809 Supplier Medchemexpress LLC Supplier No. HYB1337R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choline chloride is a quaternary ammonium salt and essential nutrient used as an analytical standard in biochemical and pharmacological research. It is provided as a high-purity crystalline solid suitable for analytical assays, receptor activation studies, and metabolic research.

  • Assay 99.17%.
  • CAS number 67-48-1.
  • Molecular weight 139.62 g/mol.
  • Chemical formula C5H14ClNO.
  • Suitable for in vitro and analytical applications.
  • Available in small reference quantities, such as 10 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.