Learn More
Selleck Chemical LLC Choline Chloride S4171-1g
SDP
Catalog No. 503109420
Encompass_Preferred
Choline chloride(Hepacholine) is a quaternary ammonium salt used as an additive for animal feed It is important as a precursor of acetylcholine as a methyl donor in various metabolic processes and in lipid metabolism
Catalog No. 50-310-9420 Supplier Selleck Chemical LLC Supplier No. S41711G
Choline chloride(Hepacholine) is a quaternary ammonium salt used as an additive for animal feed It is important as a precursor of acetylcholine as a methyl donor in various metabolic processes and in lipid metabolism