Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline chloride, 98+%

Catalog No. AAA1582822
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1582822 100 g
AAA1582836 500 g
AAA158280E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1582822 Supplier Thermo Scientific Chemicals Supplier No. A1582822
Only null left
Add to Cart

CAS: 67-48-1 | C5H14ClNO | 139.62 g/mol

Choline chloride helps in providing nourishment for pig feed and poultry feed. It acts as a food flavor enhancer. It is used as a precursor of acetylcholine and is a methyl donor in various metabolic processes and in lipid metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Choline chloride helps in providing nourishment for pig feed and poultry feed. It acts as a food flavor enhancer. It is used as a precursor of acetylcholine and is a methyl donor in various metabolic processes and in lipid metabolism.

Solubility
Soluble in water, ethanol, methanol and alcohol.

Notes
Incompatible with strong oxidizing agents, strong bases, strong acids, alkalies and alloys.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-48-1
Molecular Formula C5H14ClNO
Molecular Weight (g/mol) 139.62
MDL Number MFCD00011721
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
Synonym choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
PubChem CID 6209
ChEBI CHEBI:133341
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CCO

Specifications

Melting Point ∽305°C (decomposition)
Assay Percent Range ≥98%
Quantity 100 g
Beilstein 3563126
Sensitivity Hygroscopic
Merck Index 14,2206
Solubility Information Soluble in water,ethanol,methanol and alcohol.
Formula Weight 139.63
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Choline Chloride
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a
EINECSNumber 200-655-4
RTECSNumber KH2975000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.