Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline chloride, 98+% (dry basis)

Catalog No. AAJ13410A9
Change view
Click to view available options
Quantity:
5 lb.
10 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ13410A9 10 kg Plastic bottle
AAJ13410P5 5 lb. Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ13410A9 Supplier Thermo Scientific Supplier No. J13410A9
Only null left
Add to Cart

CAS: 67-48-1 | C5H14ClNO | 139.62 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-48-1
Molecular Formula C5H14ClNO
Molecular Weight (g/mol) 139.62
MDL Number MFCD00011721
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
Synonym choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
PubChem CID 6209
ChEBI CHEBI:133341
SMILES [Cl-].C[N+](C)(C)CCO

Specifications

Melting Point ∼305°C (decomposition)
Packaging Plastic bottle
Quantity 10 kg
Beilstein 3563126
IUPAC Name (2-hydroxyethyl)trimethylazanium chloride
Formula Weight 139.62
Percent Purity ≥98 (dry basis)
Physical Form Crystalline Powder
Chemical Name or Material Choline chloride
EINECSNumber 200-655-4
RTECSNumber KH2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.