Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline chloride, 99%, MP Biomedicals™

Catalog No. ICN10138605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10138605 5 kg
ICN10138601 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10138605 Supplier MP Biomedicals, Inc Supplier No. 0210138605
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-48-1
Molecular Formula C5H14ClNO
Molecular Weight (g/mol) 139.62
MDL Number MFCD00011721
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
Synonym choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
PubChem CID 6209
ChEBI CHEBI:133341
IUPAC Name (2-hydroxyethyl)trimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)CCO

Specifications

Color White
Density 1.1g/cm3 at 20°C
Assay Percent Range 0.99
Quantity 5 kg
Formula Weight 139.63
Physical Form Crystalline Powder
Chemical Name or Material Choline chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.