Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Chloride, FCC, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18601674 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-674 500 g Poly Bottle
18-601-675 2.5 kg Poly Bottle
18-601-676 12 kg Poly Bottle
18-601-677 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-674 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CH147500GMPL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

CH147, 67-48-1, C5H14ClNO

Spectrum™ Chemical Choline Chloride, FCC is an essential nutrient and typically grouped with B-complex vitamins. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 67-48-1
Molecular Formula C5H14ClNO
Molecular Weight (g/mol) 139.62
MDL Number MFCD00011721
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
IUPAC Name (2-hydroxyethyl)trimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)CCO

Specifications

CAS Max % 100%
Ignition Residue 0.05%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 98-100.5%
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Choline Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.