Learn More
Medchemexpress LLC Choline chloride (Standard) | 67-48-1 | 99.2% | 25 MG
SDP
Catalog No. 5000414175
Encompass_Preferred
Choline chloride (Standard) is the analytical standard of Choline (chloride) and is intended for research and analytical applications. It is an essential nutrient that activates alpha7 nicotinic receptors and possesses analgesic and anti-inflammatory activity. Glycerophosphoinositol choline, which is related to choline, can impact diseases such as liver disease, atherosclerosis, and neurological disorders.
  • Analytical standard
  • Activates alpha7 nicotinic receptors
  • Has analgesic activity
  • Has anti-inflammatory activity
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-004-14175 Supplier Medchemexpress LLC Supplier No. 25MGHYB1337R
Choline chloride (Standard) is the analytical standard of Choline (chloride) and is intended for research and analytical applications. It is an essential nutrient that activates alpha7 nicotinic receptors and possesses analgesic and anti-inflammatory activity. Glycerophosphoinositol choline, which is related to choline, can impact diseases such as liver disease, atherosclerosis, and neurological disorders.
  • Analytical standard
  • Activates alpha7 nicotinic receptors
  • Has analgesic activity
  • Has anti-inflammatory activity
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS