Learn More
Medchemexpress LLC Choline-d13 (chloride) | 352438-97-2 | MFCD01074298 | 99.9% | 152.70 g/mol | C5HD13ClNO | 10 MG
SDP
Catalog No. 5000297982
Encompass_Preferred
Choline-d13 chloride is the deuterium-labeled analog of choline chloride supplied for laboratory research use as a stable isotope tracer and analytical internal standard. It is provided as a research-grade, high-purity solid with supporting documentation for safe laboratory handling.
  • Deuterium-labeled choline chloride suitable for tracer and analytical applications.
  • High isotopic/purity level reported at 99.9%.
  • Molecular weight approximately 152.70 g/mol, CAS number 352438-97-2.
  • Available in small, research-sized quantities (10 MG vial option).
  • Manufacturer data sheet and SDS available for handling and safety information.
Catalog No. 50-002-97982 Supplier Medchemexpress LLC Supplier No. HYB1337S310MG
Choline-d13 chloride is the deuterium-labeled analog of choline chloride supplied for laboratory research use as a stable isotope tracer and analytical internal standard. It is provided as a research-grade, high-purity solid with supporting documentation for safe laboratory handling.
  • Deuterium-labeled choline chloride suitable for tracer and analytical applications.
  • High isotopic/purity level reported at 99.9%.
  • Molecular weight approximately 152.70 g/mol, CAS number 352438-97-2.
  • Available in small, research-sized quantities (10 MG vial option).
  • Manufacturer data sheet and SDS available for handling and safety information.