Learn More
Medchemexpress LLC Choline-d4 chloride | 285979-70-6 | 99.7% | 143.65 | C5H10D4ClNO | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB1337S10MG
Encompass_Preferred
Choline-d4 chloride is the deuterium-labeled form of choline chloride intended for research and analytical applications. It is a quaternary ammonium salt and a substrate for choline acetyltransferase, commonly used as an internal standard or tracer in mass spectrometry and metabolic studies. This material is supplied for research use only and is accompanied by supporting documentation for quality and safety.
  • Deuterium-labeled tracer for analytical and metabolic studies.
  • Suitable as an internal standard for LC-MS/MS quantitation.
  • Quaternary ammonium salt chemistry compatible with aqueous workflows.
  • Provided with datasheet, SDS, and COA for lot-specific information.
  • Available in small microgram/milligram pack sizes for analytical use.
Catalog No. 50-002-97796