Learn More
Medchemexpress LLC Choline-d4 chloride | 285979-70-6 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1337S1MG
Encompass_Preferred
Choline-d4 chloride is the deuterium labeled Choline chloride. It is an organic compound and a quaternary ammonium salt, an acyl group acceptor and choline acetyltransferase substrate, also an important additive in feed, especially for chickens where it accelerates growth. This product is for research use only.
  • Deuterium labeled Choline chloride
  • Tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-11865