Learn More
Medchemexpress LLC Choline-d4 (chloride) | 285979-70-6 | 99.9% | 25 MG
SDP
Catalog No. 5000413843
Encompass_Preferred
Choline-d4 (chloride) is the deuterium labeled Choline chloride, an organic compound and a quaternary ammonium salt. It functions as an acyl group acceptor and choline acetyltransferase substrate. This compound is also an important additive in feed, particularly for chickens where it accelerates growth. It is for research use only.
  • Used as a tracer for quantitation during drug development.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-13843 Supplier Medchemexpress LLC Supplier No. 25MGHYB1337S
Choline-d4 (chloride) is the deuterium labeled Choline chloride, an organic compound and a quaternary ammonium salt. It functions as an acyl group acceptor and choline acetyltransferase substrate. This compound is also an important additive in feed, particularly for chickens where it accelerates growth. It is for research use only.
  • Used as a tracer for quantitation during drug development.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles of drugs.