Learn More
Medchemexpress LLC Choline-d4 chloride | 285979-70-6 | 99.9% | 5 MG
SDP
Catalog No. 5000411573
Encompass_Preferred
Choline-d4 (chloride) is the deuterium-labeled version of Choline chloride. Choline chloride is an organic compound and a quaternary ammonium salt. It acts as an acyl group acceptor and a substrate for choline acetyltransferase. Additionally, it is an important feed additive, particularly for chickens, where it helps accelerate growth. It is intended for research use only and not for sale to patients.
  • Deuterium labeled
  • Can be used as a tracer
  • Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS
  • Stable heavy isotopes, when incorporated into drug molecules, have the potential to influence the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-11573 Supplier Medchemexpress LLC Supplier No. HYB1337S5MG
Choline-d4 (chloride) is the deuterium-labeled version of Choline chloride. Choline chloride is an organic compound and a quaternary ammonium salt. It acts as an acyl group acceptor and a substrate for choline acetyltransferase. Additionally, it is an important feed additive, particularly for chickens, where it helps accelerate growth. It is intended for research use only and not for sale to patients.
  • Deuterium labeled
  • Can be used as a tracer
  • Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS
  • Stable heavy isotopes, when incorporated into drug molecules, have the potential to influence the pharmacokinetic and metabolic profiles of drugs