Learn More
Medchemexpress LLC Choline-d9 chloride | 61037-86-3 | 99.9% | 50 MG
SDP
Catalog No. 5000412065
Encompass_Preferred
Choline-d9 (chloride) is the deuterium-labeled form of choline chloride, an organic compound. It functions as a quaternary ammonium salt, an acyl group acceptor, and a choline acetyltransferase substrate. Choline chloride is also utilized as an important feed additive, particularly for chickens, where it promotes growth.
  • Used as a tracer.
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can influence the pharmacokinetic and metabolic profiles of pharmaceuticals.
Catalog No. 50-004-12065 Supplier Medchemexpress LLC Supplier No. HYB1337S150MG
Choline-d9 (chloride) is the deuterium-labeled form of choline chloride, an organic compound. It functions as a quaternary ammonium salt, an acyl group acceptor, and a choline acetyltransferase substrate. Choline chloride is also utilized as an important feed additive, particularly for chickens, where it promotes growth.
  • Used as a tracer.
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can influence the pharmacokinetic and metabolic profiles of pharmaceuticals.