Learn More
Medchemexpress LLC HY-14739 10mg Medchemexpress, Choline Fenofibrate CAS:856676-23-8 Purity:>98%
SDP
Catalog No. 502081634
Encompass_Preferred
Medchemexpress, HY-14739 10mg Choline Fenofibrate CAS:856676-23-8 Choline Fenofibrate (ABT-335) is the choline salt of Fenofibric acid (HY-B0760), which releases free fenofibric acid in the gastrointestinal tract. Fenofibric acid is a PPAR activator with antihyperlipidemic effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1634 Supplier Medchemexpress LLC Supplier No. HY1473910MG
Medchemexpress, HY-14739 10mg Choline Fenofibrate CAS:856676-23-8 Choline Fenofibrate (ABT-335) is the choline salt of Fenofibric acid (HY-B0760), which releases free fenofibric acid in the gastrointestinal tract. Fenofibric acid is a PPAR activator with antihyperlipidemic effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.