Learn More
Selleck Chemical LLC Choline Fenofibrate
SDP
Catalog No. 5000003493
Encompass_Preferred
Choline fenofibrate(ABT-335) is a newly developed choline salt of fenofibric acid a synthetic phenoxy-isobutyric acid derivate with antihyperlipidemic activity that acts as an PPAR agonist
Catalog No. 50-000-03493 Supplier Selleck Chemical LLC Supplier No. S581210MG
Choline fenofibrate(ABT-335) is a newly developed choline salt of fenofibric acid a synthetic phenoxy-isobutyric acid derivate with antihyperlipidemic activity that acts as an PPAR agonist