Learn More
Medchemexpress LLC Choline Fenofibrate | 856676-23-8 | 99.5% | 421.91 | 25 MG
SDP
Catalog No. 5000363826
Encompass_Preferred
Choline Fenofibrate (ABT-335) is a choline salt of Fenofibric acid that releases free Fenofibric acid in the gastrointestinal tract. Fenofibric acid acts as a PPAR activator with an antihyperlipidemic effect. It is for research use only.
  • PPAR activator
  • Antihyperlipidemic effect
  • Excellent absolute oral bioavailability (rat 93.4%)
  • Terminal elimination half-life (rat 5.4 h)
Catalog No. 50-003-63826 Supplier Medchemexpress LLC Supplier No. 25MGHY14739
Choline Fenofibrate (ABT-335) is a choline salt of Fenofibric acid that releases free Fenofibric acid in the gastrointestinal tract. Fenofibric acid acts as a PPAR activator with an antihyperlipidemic effect. It is for research use only.
  • PPAR activator
  • Antihyperlipidemic effect
  • Excellent absolute oral bioavailability (rat 93.4%)
  • Terminal elimination half-life (rat 5.4 h)