Learn More
Medchemexpress LLC Choline Fenofibrate | 856676-23-8 | 99.5% | 421.91 | 50 MG
SDP
Catalog No. 5000362324
Encompass_Preferred
Choline Fenofibrate is a choline salt of Fenofibric acid. It releases free fenofibric acid in the gastrointestinal tract, acting as a PPAR activator with an antihyperlipidemic effect. This product is for research use only.
  • Choline salt of fenofibric acid
  • Releases free fenofibric acid
  • PPAR activator
  • Antihyperlipidemic effect
  • For research use only
  • Excellent absolute oral bioavailability (rat 93.4%)
  • High Cmax (rat 7944.2 ng/mL)
  • Terminal elimination half-life (rat 5.4 h)
Catalog No. 50-003-62324 Supplier Medchemexpress LLC Supplier No. HY1473950MG
Choline Fenofibrate is a choline salt of Fenofibric acid. It releases free fenofibric acid in the gastrointestinal tract, acting as a PPAR activator with an antihyperlipidemic effect. This product is for research use only.
  • Choline salt of fenofibric acid
  • Releases free fenofibric acid
  • PPAR activator
  • Antihyperlipidemic effect
  • For research use only
  • Excellent absolute oral bioavailability (rat 93.4%)
  • High Cmax (rat 7944.2 ng/mL)
  • Terminal elimination half-life (rat 5.4 h)