Learn More
Selleck Chemical LLC Choline Glycerophosphate S4972-1g
SDP
Catalog No. 503109006
Encompass_Preferred
Choline Glycerophosphate (Glycerophosphorylcholine) is a choline derivative and one of the two major forms of choline storage (along with phosphocholine) in the cytosol
Catalog No. 50-310-9006 Supplier Selleck Chemical LLC Supplier No. S49721G
Choline Glycerophosphate (Glycerophosphorylcholine) is a choline derivative and one of the two major forms of choline storage (along with phosphocholine) in the cytosol