Learn More
Selleck Chemical LLC Choline Glycerophosphate S4972-25mg
SDP
Catalog No. 503110868
Encompass_Preferred
Choline Glycerophosphate (Glycerophosphorylcholine) is a choline derivative and one of the two major forms of choline storage (along with phosphocholine) in the cytosol
Catalog No. 50-311-0868 Supplier Selleck Chemical LLC Supplier No. S497225MG
Choline Glycerophosphate (Glycerophosphorylcholine) is a choline derivative and one of the two major forms of choline storage (along with phosphocholine) in the cytosol