Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Iodide 98.0+%, TCI America™
SDP

Catalog No. C257325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C257325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C257325G Supplier TCI America Supplier No. C257325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17773-10-3
Molecular Formula C5H14INO
Molecular Weight (g/mol) 231.08
MDL Number MFCD00011900
InChI Key FNPBHXSBDADRBT-UHFFFAOYSA-M
Synonym choline iodide, jodoetano, dilatol zambon, 2-hydroxy-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, iodide, 2-hydroxyethyl trimethylammonium iodide, cholineiodide, acmc-209ed9, d0q2de, 2-hydroxyethyl trimethyl azanium iodide
PubChem CID 87300
IUPAC Name (2-hydroxyethyl)trimethylazanium iodide
SMILES [I-].C[N+](C)(C)CCO

Specifications

Melting Point 264°C
Color White-Yellow
Quantity 25 g
Formula Weight 231.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Choline Iodide
EINECSNumber (2)-0341&(9)-1994
RTECSNumber GA4100000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.