Learn More
Medchemexpress LLC Choline tosylate 25g
SDP
Catalog No. 5000206135
Encompass_Preferred
Choline tosylate is a nucleophilic compound that inhibits phospholipase A2 and phospholipase C Choline tosylate inhibits tumor growth in mice by inhibiting the formation of diacylglycerol (DAG)[1][2]
Catalog No. 50-002-06135 Supplier Medchemexpress LLC Supplier No. 25GHYW100403
Choline tosylate is a nucleophilic compound that inhibits phospholipase A2 and phospholipase C Choline tosylate inhibits tumor growth in mice by inhibiting the formation of diacylglycerol (DAG)[1][2]