Learn More
Medchemexpress LLC Choline tosylate 500g
SDP
Catalog No. 5000206201
Encompass_Preferred
Choline tosylate is a nucleophilic compound that inhibits phospholipase A2 and phospholipase C Choline tosylate inhibits tumor growth in mice by inhibiting the formation of diacylglycerol (DAG)[1][2]
Catalog No. 50-002-06201 Supplier Medchemexpress LLC Supplier No. HYW100403500G
Choline tosylate is a nucleophilic compound that inhibits phospholipase A2 and phospholipase C Choline tosylate inhibits tumor growth in mice by inhibiting the formation of diacylglycerol (DAG)[1][2]