Learn More
SPEX CERTIPREP LLC 3P Solutions Chlorine Snips
Catalog No. NC0488881
Encompass_Preferred
(NSI by Zeptometrix) A box of 25 SNIPs for use in verifying the accuracy of your ultra low residual chlorine analysis. Diluting contents of one SNIP to 1000 mL yields a 0.100 mg/L sample. No pipets required, just snip and pour. Each box yields 25 QA analyses.
Catalog No. NC0488881 Supplier SPEX CERTIPREP LLC 3P Supplier No. QCI118
(NSI by Zeptometrix) A box of 25 SNIPs for use in verifying the accuracy of your ultra low residual chlorine analysis. Diluting contents of one SNIP to 1000 mL yields a 0.100 mg/L sample. No pipets required, just snip and pour. Each box yields 25 QA analyses.