Learn More
Cayman Chemical Cholyl-1 3-dIamInopropn 10mg
SDP
Catalog No. 502945465
Encompass_Preferred
A polyamine bile amidate; has been used as a standard for the detection of endogenous bile acid amidates in mammalian intestinal and fecal samples
Catalog No. 50-294-5465 Supplier Cayman Chemical Supplier No. 4089810 MG
A polyamine bile amidate; has been used as a standard for the detection of endogenous bile acid amidates in mammalian intestinal and fecal samples