Learn More
Cayman Chemical Cholyl-cadaverIn 10mg
SDP
Catalog No. 502944855
Encompass_Preferred
A polyamine bile amidate; has been used as a standard for the detection of endogenous bile acid amidates in mammalian fecal samples
Catalog No. 50-294-4855 Supplier Cayman Chemical Supplier No. 4089710 MG
A polyamine bile amidate; has been used as a standard for the detection of endogenous bile acid amidates in mammalian fecal samples