Learn More
Promocell GmbH PromoCell Chondrocyte Basal Medium 500 ml
Catalog No. 50306265
Encompass
PromoCell Chondrocyte Growth Medium has been developed for the in vitro cultivation of human chondrocytes. The fetal calf serum in the medium is strictly pretested for optimal performance.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for primary porcine and rat chondrocytes.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.1 ml / ml |
Catalog No. 50-306-265 Supplier Promocell GmbH Supplier No. C27111
PromoCell Chondrocyte Growth Medium has been developed for the in vitro cultivation of human chondrocytes. The fetal calf serum in the medium is strictly pretested for optimal performance.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for primary porcine and rat chondrocytes.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.1 ml / ml |