Learn More
ATCC Chondrocyte Differentiation Tool
Catalog No. 502383031
Encompass
The Chondrocyte Differentiation Tool is a complete differentiation medium designed to induce chondrogenesis in actively proliferating Adipose-derived Mesenchymal Stem Cells with high efficiency. The Chondrocyte Differentiation Tool provides enough medium to differentiate up to ~ 13.5 x 10^7 cells, sufficient to seed 20 wells of a 48-well tissue culture plate when using alginate encapsulation.
Catalog No. 50-238-3031 Supplier ATCC Supplier No. PCS500051
The Chondrocyte Differentiation Tool is a complete differentiation medium designed to induce chondrogenesis in actively proliferating Adipose-derived Mesenchymal Stem Cells with high efficiency. The Chondrocyte Differentiation Tool provides enough medium to differentiate up to ~ 13.5 x 10^7 cells, sufficient to seed 20 wells of a 48-well tissue culture plate when using alginate encapsulation.