Learn More
Lonza Walkersville Chondrocyte ReagentPack Subculture Reagents
Catalog No. NC1605690
Encompass
Chondrocyte ReagentPack Subculture Kits have been designed specifically for the passaging of primary chondrocytes.Each kit contains 100 ml each of: Trypsin/EDTA Trypsin Neutralizing Solution HEPES Buffered SalineThe Chondrocyte ReagentPack Subculture Kit is recommended for use with Lonza's primary chondrocytes for effective trypsinization and cell passaging process.Content:1 x Bottle Trypsin/EDTA 100 mL1 x Bottle Trypsin Neutralizing Solution 100 mL1x Bottle HEPES Buffered Saline Solution 100 mL
This item is not returnable. View return policy
Catalog No. NC1605690 Supplier Lonza Walkersville Supplier No. CC3233
Chondrocyte ReagentPack Subculture Kits have been designed specifically for the passaging of primary chondrocytes.Each kit contains 100 ml each of: Trypsin/EDTA Trypsin Neutralizing Solution HEPES Buffered SalineThe Chondrocyte ReagentPack Subculture Kit is recommended for use with Lonza's primary chondrocytes for effective trypsinization and cell passaging process.Content:1 x Bottle Trypsin/EDTA 100 mL1 x Bottle Trypsin Neutralizing Solution 100 mL1x Bottle HEPES Buffered Saline Solution 100 mL