Learn More
Lonza Walkersville Human Mesenchymal Stem Cell (hMSC) Chondrogenic Differentiation Medium BulletKit
Catalog No. NC9986228
Encompass
Stem cells possess the unique ability to differentiate into multiple lineages of cells including chondrocytes. Lonza’s hMSC Chondrogenic Differentiation BulletKit Medium is a serum containing medium designed to induce chondrogenic differentiation, with the addition of TGF-β3, of human bone marrow derived mesenchymal stem cells into mature, functionally active chondrocytes in as few as 14 days. Differentiation can then be validated by a variety of methods including staining with alcian blue or safranin.Partial List of Cells Differentiated with hMSC Chondrogenic Differentiation BulletKit Medium*:Human bone marrow derived stem cellsHuman umbilical cord blood derived stem cellsHuman umbilical cord matrix derived stem cellsC57BL/6 mice bone marrow derived stem cellshuman adipose derived stem cellsHuman hamstring tendon derived stem cellsContent:1 x hMSC Chondrogenic Basal Medium (PT-3925)1 x hMSC Chondrogenic SingleQuot Kit (PT-4121)
Catalog No. NC9986228 Supplier Lonza Walkersville Supplier No. PT3003
Stem cells possess the unique ability to differentiate into multiple lineages of cells including chondrocytes. Lonza’s hMSC Chondrogenic Differentiation BulletKit Medium is a serum containing medium designed to induce chondrogenic differentiation, with the addition of TGF-β3, of human bone marrow derived mesenchymal stem cells into mature, functionally active chondrocytes in as few as 14 days. Differentiation can then be validated by a variety of methods including staining with alcian blue or safranin.Partial List of Cells Differentiated with hMSC Chondrogenic Differentiation BulletKit Medium*:Human bone marrow derived stem cellsHuman umbilical cord blood derived stem cellsHuman umbilical cord matrix derived stem cellsC57BL/6 mice bone marrow derived stem cellshuman adipose derived stem cellsHuman hamstring tendon derived stem cellsContent:1 x hMSC Chondrogenic Basal Medium (PT-3925)1 x hMSC Chondrogenic SingleQuot Kit (PT-4121)