Learn More
Cayman Chemical Chondroitin Sulfate (sodium salt), CAS: 9082-07-9, 10 g
SDP
Catalog No. 502918203
Encompass_Preferred
A glycosaminoglycan; contains a GAG-protein linker region and repeating disaccharide subunits composed of GlcA and GalNAc; chains of various lengths covalently attach to proteins to form CSPGs
Catalog No. 50-291-8203 Supplier Cayman Chemical Supplier No. 3780110 G
A glycosaminoglycan; contains a GAG-protein linker region and repeating disaccharide subunits composed of GlcA and GalNAc; chains of various lengths covalently attach to proteins to form CSPGs