Learn More
Apexbio Technology LLC Chondroitin sulfate 9007-28-7 1g
SDP
Catalog No. 5000090129
Encompass_Preferred
Chondroitin sulfate (9007-28-7) is a glycosaminoglycan designed to modulate inflammatory and apoptotic processes thereby regulating extracellular matrix homeostasis Chondroitin sulfate exerts its biological activity primarily by inhibiting inflammatory mediators and reducing cell apoptosis through interactions with various biological molecules Based on these pharmacological properties chondroitin sulfate holds research potential in investigating cartilage degradation inflammatory response mechanisms and therapeutic targets related to joint degeneration and tissue remodeling
Catalog No. 50-000-90129 Supplier Apexbio Technology LLC Supplier No. N27301000
Chondroitin sulfate (9007-28-7) is a glycosaminoglycan designed to modulate inflammatory and apoptotic processes thereby regulating extracellular matrix homeostasis Chondroitin sulfate exerts its biological activity primarily by inhibiting inflammatory mediators and reducing cell apoptosis through interactions with various biological molecules Based on these pharmacological properties chondroitin sulfate holds research potential in investigating cartilage degradation inflammatory response mechanisms and therapeutic targets related to joint degeneration and tissue remodeling